Anti-KAT8 Rabbit Polyclonal Antibody

Supplier: BioVision
6149-100
10008-232EA 391.11 USD
10008-232
Anti-KAT8 Rabbit Polyclonal Antibody
This gene encodes a member of the MYST histone acetylase protein family. The encoded protein has a characteristic MYST domain containing an acetyl­CoA­binding site, a chromodomain typical of proteins which bind histones, and a C2HC­type zinc finger. Multiple transcript variants encoding different isoforms have been found for this gene. [Provided by RefSeq, Feb 2012].

Recommended Dilutions: Western Blot: 1:500-1:1000

Type: Primary
Antigen: KAT8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR