Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CBX5 Rabbit Polyclonal Antibody

Supplier: BioVision

6148-100
10008-230EA 345.05 USD InStock
10008-230
Anti-CBX5 Rabbit Polyclonal Antibody
Antibodies
Heterochromatin protein 1 (HP1) is a family of heterochromatic adaptor molecules involved in both gene silencing and higher order chromatin structure. All three HP1 family members (α, β, and γ) are primarily associated with centromeric heterochromatin; However, HP1β and γ also localize to euchromatic sites in the genome. HP1 proteins are approximately 25 kDa in size and contain a conserved amino terminal chromodomain, followed by a variable hinge region and a conserved carboxy­terminal chromoshadow domain. The chromodomain facilitates binding to histone H3 tri­methylated at Lys9, a histone "mark" closely associated with centromeric heterochromatin. The variable hinge region binds both RNA and DNA in a sequence independent manner. The chromoshadow domain mediates the dimerization of HP1 proteins, in addition to binding multiple proteins  implicated in gene silencing and heterochromatin  formation, including the SUV39H histone methyltransferase, the DNMT1 and DNMT3a DNA  methyltransferases, and the p150 subunit of chromatin­assembly factor­1 (CAF1). In addition to contributing to heterochromatin formation and propagation, HP1 and SUV39H are also found complexed with retinoblastoma (Rb) and E2F6 proteins, both of which function to repress euchromatic gene transcription in quiescent cells. HP1 proteins are subject to multiple types of post­translational modifications, including phosphorylation, acetylation, methylation, ubiquitination, and sumoylation, suggesting multiple means of regulation.

Recommended Dilutions: Western Blot: 1:500-1:2000, IHC: 1:50-1:200

Type: Primary
Antigen: CBX5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty