Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-UHRF2 Rabbit Polyclonal Antibody

Supplier: BioVision

6145-100
10008-224EA 345.05 USD InStock
10008-224
Anti-UHRF2 Rabbit Polyclonal Antibody
Antibodies
This gene encodes a nuclear protein which is involved in cell cycle regulation. The encoded protein is a ubiquitin ligase capable of ubiquinating PCNP (PEST­containing nuclear protein), and together they may play a role in tumorigenesis. The encoded protein contains an NIRF_N domain, a PHD finger, a set­ and ring associated (SRA) domain, and a RING finger domain and several of these domains have been shown to be essential for the regulation of cell proliferation. This protein may also have a role in intranuclear degradation of polyglutamine aggregates. Alternative splicing results in multiple transcript variants, some of which are non­protein coding. [Provided by RefSeq, Feb 2012].

Recommended Dilutions: Western Blot: 1:500-1:2000, IHC: 1:50-1:200, Immunofluorescence: 1:20-1:50.

Type: Primary
Antigen: UHRF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty