Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-UHRF1 Rabbit Polyclonal Antibody

Supplier: BioVision

6144-100
10008-222EA 345.05 USD InStock
10008-222
Anti-UHRF1 Rabbit Polyclonal Antibody
Antibodies
UHRF1, also named as ICBP90, NP95 and RNF106, is a putative E3 ubiquitin protein ligase. It may participate in methylation dependent transcriptional regulation. UHRF1 binds to inverted 5'­CCAAT­3' box 2 in the TOP2A promoter, and activates TOP2A expression. It is important for G1/S transition and may be involved in DNA repair and chromosomal stability. UHRF1's ability to repress its direct target gene expression is dependent on PHD (UHRF1) binding to unmodified H3R2.

Recommended Dilutions: Western Blot: 1:500-1:2000, IHC: 1:50-1:200, Immunofluorescence: 1:20-1:50, IP: 1:20-1:50, CHIP: 1:20-1:50

Type: Primary
Antigen: UHRF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty