Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-USP2 Rabbit Polyclonal Antibody

Supplier: BioVision

6141-100
10008-216EA 345.05 USD InStock
10008-216
Anti-USP2 Rabbit Polyclonal Antibody
Antibodies
Ubiquitinating enzymes (UBEs) catalyze protein ubiquitination, a reversible process countered by deubiquitinating enzymes (DUB) action. There are five DUB subfamilies including the USP, UCH, OTU, MJD, and JAMM enzymes. Ubiquitin­specific­processing protease 2 (USP2) belongs to the USP (UBP/UCH type 2) subfamily and is characterized by its C19 peptidase activity, which is involved in ubiquitin recycling and in the disassembly of various forms of polymeric ubiquitin and ubiquitin like protein complexes. There is mounting evidence that USP2 functions as an oncoprotein through multiple mechanisms. In human prostate cancer, USP2 is highly overexpressed and drives tumor growth by binding to and stabilizing fatty acid synthase through deubiquitination. It has also been demonstrated that USP2 can bind and deubiquitinate both Mdm2 and cyclin D1, which leads to their stabilization and enhanced cell proliferation.

Recommended Dilutions: Western Blot: 1:200-1:500 dilution, IHC: 1:50 - 1:100 dilution. Immunofluorescence: 1:20 - 1:50 dilution

Type: Primary
Antigen: USP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty