Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-USP1 Rabbit Polyclonal Antibody

Supplier: BioVision

6140-100
10008-214EA 345.05 USD InStock
10008-214
Anti-USP1 Rabbit Polyclonal Antibody
Antibodies
Ubiquitinating enzymes (UBEs) catalyze protein ubiquitination, a reversible process countered by deubiquitinating enzymes (DUB) action. There are five DUB subfamilies including the USP, UCH, OTU, MJD, and JAMM enzymes. USP1 belongs to the USP subfamily and is regulated in a cell cycle dependent manner by both transcriptional and ubiquitin­proteosomal mechanisms. USP1 is a nuclear protein and localizes to chromatin where it is specifically associated with FANCD2. USP1 deubiquitinates monoubiquitinated FANCD2, which plays an important role in DNA damage repair and Chk1 protein stability. USP1 deubiquitinates monoubiquitinated PCNA, thereby negatively regulating PCNA­mediated translation synthesis (TLS) during DNA repair. USP1 interaction with UAF1, a WD40 repeat containing protein, leads to formation of an activated USP1/UAF1 complex, which is required for the deubiquitinase activity of USP1.

Recommended Dilutions: Western Blot: 1:200-1:500 dilution, IHC: 1:50 - 1:100 dilution

Type: Primary
Antigen: USP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty