Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SIRT2 Rabbit Polyclonal Antibody

Supplier: BioVision

6138-100
10008-212EA 345.05 USD InStock
10008-212
Anti-SIRT2 Rabbit Polyclonal Antibody
Antibodies
The sirtuins represent a distinct class of trichostatin A-insensitive lysyl-deacetylases (class III HDACs) and have been shown to catalyze a reaction that couples lysine deacetylation to the formation of nicotinamide and O-acetyl-ADP-ribose from NAD+ and the abstracted acetyl group. This SIRT family of genes is highly conserved from prokaryotes to eukaryotes and is involved in diverse processes including transcriptional regulation, cell cycle progression, DNA damage repair and aging. SIRT2, a human homolog of the yeast SIR2 (silent information regulator­2), functions as transcriptional silencing mediator at mating­type loci, telomeres and ribosomal gene clusters. SIRT2 expression increases dramatically during mitosis and is multiply phosphorylated at the G (2)/M transition of the cell cycle. SIRT2 is part of a phosphorylation cascade where it is phosphorylated late in G (2), during M, and into the period of cytokinesis. Inhibition of SIRT2 is reported to rescue alpha­synuclein toxicity and modify inclusion morphology in a cellular model of Parkinson's disease.

  • Recommended Dilutions: Western Blot: 1:500-1:2000 dilution, IHC: 1:50 - 1:200 dilution

Type: Primary
Antigen: SIRT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty