Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-USP34 Chicken Polyclonal Antibody

Supplier: BioVision

6136-50
10008-208EA 149.35 USD InStock
10008-208
Anti-USP34 Chicken Polyclonal Antibody
Antibodies
USP34 Core is a ubiquitin hydrolase that can remove conjugated ubiquitin from AXIN1 and AXIN2, thereby acting as a regulator of Wnt signaling pathway. It acts as an activator of the Wnt signaling pathway downstream of the beta-catenin destruction complex by deubiquitinating and stabilizing AXIN1 and AXIN2, leading to promote nuclear accumulation of AXIN1 and AXIN2 and positively regulate beta-catenin (CTNBB1)-mediated transcription. It recognizes and hydrolyzes the peptide bond at the C-terminal Gly of ubiquitin. It is involved in the processing of poly-ubiquitin precursors as well as that of ubiquinated proteins. Its catalytic core consists of residues 1892-2241.

Recommended Dilutions: Western Blot: Robust detection of 100 ng of recombinant protein was possible when antibody was used at a final concentration of 5 μg/mL

Type: Primary
Antigen: USP34
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty