Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-USP15 Chicken Polyclonal Antibody

Supplier: BioVision

6135-50
10008-206EA 149.35 USD InStock
10008-206
Anti-USP15 Chicken Polyclonal Antibody
Antibodies
The ubiquitin (Ub) pathway involves three sequential enzymatic steps that facilitate the conjugation of Ub and Ub-like molecules to specific protein substrates. Through the use of a wide range of enzymes that can add or remove ubiquitin, the Ub pathway controls many intracellular processes such as signal transduction, transcriptional activation and cell cycle progression. USP15 (ubiquitin specific peptidase 15), also known as UNPH4, is a member of the peptidase C19 family of proteins. Expressed in kidney, liver, placenta, ovary, lung, thymus, heart and skeletal muscle, USP15 localizes to the cytoplasm and the nucleus, contains one DUSP domain and functions as a deubiquitinating enzyme that cleaves ubiquitin residues from both ubiquitinylated proteins and ubiquitin-fused precursors, thereby saving these proteins from proteasomal degradation. Via its DUSP domain, USP15 plays a role in the regulation of the COP9 signalosome (CSN) complex. Three isoforms exist for USP15 due to alternative splicing events.

Recommended Dilutions: Western Blot: Robust detection of 100 ng of recombinant protein was possible when antibody was used at a final concentration of 5 μg/mL

Type: Primary
Antigen: USP15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty