Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-USP5 Chicken Polyclonal Antibody

Supplier: BioVision

6132-50
10008-200EA 149.35 USD InStock
10008-200
Anti-USP5 Chicken Polyclonal Antibody
Antibodies
The ubiquitin (Ub) pathway involves three sequential enzymatic steps that facilitate the conjugation of Ub and Ub-like molecules to specific protein substrates. Through the use of a wide range of enzymes that can add or remove ubiquitin, the Ub pathway controls many intracellular processes such as signal transduction, transcriptional activation and cell cycle progression. USP5 (ubiquitin specific peptidase 5), also known as ISOT (isopeptidase T), is a 858 amino acid zinc-binding deubiquitinating enzyme that participates in the Ub pathway. A member of the peptidase C19 family, the catalytic activity of USP5 involves a combination of the ubiquitin carboxyl-terminal thiolester and water to produce ubiquitin and a thiol. USP5 contains two UBA domains and one UBP-type zinc finger. USP5 is responsible for disassembling unanchored polyubiquitin chains in the cell. Full length USP5 mediated cleavage of Ub-PLA2 is significantly enhanced is enhanced by low concentrations of mono ubiquitin.

Recommended Dilutions: Western Blot: Robust detection of 100 ng of recombinant protein was possible when antibody was used at a final concentration of 5 μg/mL

Type: Primary
Antigen: USP5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty