Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TAOK1 Rabbit Polyclonal Antibody

Supplier: BioVision

3776-100
10004-558EA 272.95 USD InStock
10004-558
Anti-TAOK1 Rabbit Polyclonal Antibody
Antibodies
Serine/threonine-protein kinase TAO1 is an enzyme that in humans is encoded by the TAOK1 gene.

Recommended Dilutions: Western Blot: 1:500-1:2000

Type: Primary
Antigen: TAOK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty