Anti-TRIP1 Rabbit Polyclonal Antibody

Supplier: BioVision
3775-100
10004-556EA 286.04 USD
10004-556
Anti-TRIP1 Rabbit Polyclonal Antibody
Thyroid receptor interacting protein 1 (TRIP1) was originally identified as a protein that interacts with the thyroid receptor and is homologous to the yeast SUG1 protein, a transcriptional coactivator. It was later identified as an ATPase subunit of the 26S proteasome, a multicatalytic proteinase complex, and has also been shown to associate with nuclear receptors and transcription factors. TRIP1/SUG1 may function to regulate transcription factor abundance by targeting them for proteasome degradation.

Recommended Dilutions: Western Blot: 1:500-1:2500

Type: Primary
Antigen: TRIP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR