Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TIF1a Rabbit Polyclonal Antibody

Supplier: BioVision

3774-100
10004-554EA 252.35 USD InStock
10004-554
Anti-TIF1a Rabbit Polyclonal Antibody
Antibodies
TIF1 Alpha/TRIM24 is a member of a family of proteins that control transcription and chromatin remodeling through their interaction with transcription factors. The family of TIF1s (transcriptional intermediary factor 1) includes TIF1-alpha, TIF1-beta, and TIF1-gamma. TIF1-alpha plays an important role in development and mediates transcriptional control by interacting with the AF2 region of nuclear receptors such as estrogen, retinoic acid, and vitamin D3 receptors. TIF1-alpha has been shown to be a repressor of transcription. This transcriptional repression involves histone deacetylation and interaction with members of the heterochromatin protein 1 family.

Recommended Dilutions: Western Blot: 1:500-1:2000

Type: Primary
Antigen: TIF1a
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty