Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SRC3 Rabbit Polyclonal Antibody

Supplier: BioVision

3773-100
10004-552EA 252.35 USD InStock
10004-552
Anti-SRC3 Rabbit Polyclonal Antibody
Antibodies
Steroid receptor coactivator 3 (SRC3) belongs to the SRC family of proteins that interacts with nuclear receptors to facilitate transcription. The 3 homologous members of this family are SRC-1 (NCoA-1), SRC-2 (GRIP1, TIF2, or NcoA-2), and SRC-3 (p/CIP, RAC3, ACTR, AIB1, or TRAM-1). SRC3 is an oncogene that is reported to be amplified and/or overexpressed in breast and gastric cancer and stimulates AKT signaling and cell growth and tumorigenesis.

Recommended Dilutions: Western Blot: 1:500-1:2500

Type: Primary
Antigen: SRC3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty