Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ASH2 Rabbit Polyclonal Antibody

Supplier: BioVision

3772-100
10004-742EA 272.95 USD InStock
10004-742
Anti-ASH2 Rabbit Polyclonal Antibody
Antibodies
ASH2 (absent, small, or homeotic discs 2) is a subunit of a human Set1-like histone methyltransferase (HMT) complex. The Set-1 like complexes are responsible for mono-, di-, and tri-methylation of histone H3 K4. ASH2 associates with a multitude of proteins in a number of Set1-like complexes which include various combinations of MLL, MLL2, MLL3, MLL4, RBBP5,WDR5, hDPY-30, hSwd2, CXXC1, HCF1, Menin, PTIP, PA1, NCOA6, and UTX.

Recommended Dilutions: Western Blot: 1:500-1:2500

Type: Primary
Antigen: ASH2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty