Anti-FUS Rabbit Polyclonal Antibody

Supplier: BioVision
3771-100
10004-740EA 286.04 USD
10004-740
Anti-FUS Rabbit Polyclonal Antibody
FUS has been identified as a frequent translocation fusion partner of various transcription factors. FUS fusion genes have been shown to be associated with multiple tumor types which include liposarcoma, leukemia, histocytoma, and sarcoma. FUS is a multifunctional RNA-binding protein that associates with the nuclear matrix and Cajal bodies and appears to play a role in splicesome assembly, pre-mRNA splicing, DNA repair, transcriptional regulation and homologous recombination.

Recommended Dilutions: Western Blot: 1:500-1:2000

Type: Primary
Antigen: FUS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR