Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CLSPN Rabbit Polyclonal Antibody

Supplier: BioVision

3770-100
10004-738EA 252.35 USD InStock
10004-738
Anti-CLSPN Rabbit Polyclonal Antibody
Antibodies
Claspin is a checkpoint protein that interacts with and regulates checkpoint kinase 1 (Chk1) in response to replication stress. The interaction between claspin and Chk1 has been demonstrated to be required for ATR-dependent Chk1 activation. The interaction involves the phosphorylation of claspin that may be accomplished by Chk1 kinase activity. Claspin has been found to localize to chromatin in both the presence and absence of damaged DNA and to associate with PCNA, as an essential component of the DNA replication machinery. This association with the replication machinery may indicate a role for claspin in linking replication to DNA damage signaling and Chk1 activation.

Recommended Dilutions: Western Blot: 1:500-1:2000

Type: Primary
Antigen: CLSPN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty