Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ATR Rabbit Polyclonal Antibody

Supplier: BioVision

3767-100
10004-732EA 272.95 USD InStock
10004-732
Anti-ATR Rabbit Polyclonal Antibody
Antibodies
ATR (ATM and Rad3 related) is closely related to ATM (Ataxia telangiectasia, mutated) and is a member of the phosphatidylinositol 3 kinase (PI-3) family that is an early sensor of DNA damage. ATR is a serine-threonine kinase that reacts to UV damage and interruptions in replication. ATR may be able to sense DNA damage through interaction with Rad17 and 1as well as components of nucleosome remodeling complexes. In response to DNA damage, ATR has been shown to phosphorylate a multitude of substrates which include BRCA1, p53, Chk2, Rad 17, and E2F transcription factor 1.

Recommended Dilutions: Western Blot: 1:500-1:2000

Type: Primary
Antigen: ATR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty