Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MAD1 Rabbit Polyclonal Antibody

Supplier: BioVision

3765-100
10004-728EA 252.35 USD InStock
10004-728
Anti-MAD1 Rabbit Polyclonal Antibody
Antibodies
Cell cycle progression is subject to arrest at the mitotic spindle assembly checkpoint in response to incorrect spindle fiber assembly. MAD1 and MAD2 (for mitotic arrest-deficient 1 and 2) are components of the mitotic spindle checkpoint. Incorrect spindle assembly in normal cells leads to mitotic arrest. MAD1 prevents the onset of anaphase until all chromosomes are aligned correctly at the metaphase plate and is crucial for anchoring MAD2L1 to the nuclear periphery. It also plays an important role in septum positioning. MAD1 can form a homo-dimer, but may also form a heterodimer with MAD2 to form the tetrameric MAD1L1-MAD2L1 core complex. MAD1 localizes primarily to the nucleus, but during mitosis, it moves from a nuclear distribution to the centrosome, to the spindle midzone and then on to the midbody. MAD1 activity is induced by BUB1 and the protein is hyperphosphorylated after mitotic spindle damage and/or in late S through M phase. Defects in the gene encoding for MAD1, MAD1L1, play a major role in the development and progression of various cancer types.

Recommended Dilutions: Western Blot: 1:500-1:2000

Type: Primary
Antigen: MAD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty