Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-KAP-1 Rabbit Polyclonal Antibody

Supplier: BioVision

3764-100
10004-726EA 252.35 USD InStock
10004-726
Anti-KAP-1 Rabbit Polyclonal Antibody
Antibodies
KAP-1 (KRAB-Associated Protein 1) is a member of the tripartite motif RBCC/Trim domain family that contains a RING finger domain, B boxes, and an alpha helical coiled coil region. KAP-1 functions as a corepressor by interacting with the KRAB domain of KRAB zinc-finger transcriptional repressors. KAP-1 also appears to function as a scaffold for chromatin-remodeling complexes involved in transcriptional repression.

Recommended Dilutions: Western Blot: 1:500-1:2500

Type: Primary
Antigen: KAP-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty