Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HEC1 Rabbit Polyclonal Antibody

Supplier: BioVision

3763-100
10004-724EA 252.35 USD InStock
10004-724
Anti-HEC1 Rabbit Polyclonal Antibody
Antibodies
Highly expressed in cancer (Hec1) is a coiled-coil-enriched protein expressed abundantly in the S and M phases of rapidly dividing cells where it localizes to the kinetochores. Hec1 is involved in spindle checkpoint signaling. Hec1 is not expressed in terminal differentiated cells. Hec1 is expressed in tissues with high mitotic rates including testis, spleen and thymus. Hec1 is also found in the late S to M phases of bladder carcinoma cells. In dividing cells, Hec1 is required for the recruitment of Mps1 kinase and MAD1/MAD2 complexes to the kinetochores. The phosphorylation of Hec1 on Serine 165 by Nek2 is essential for faithful chromosome segregation. The binding of retinoblastoma protein to Hec1 also increases the fidelity of chromosomal segregation.

Recommended Dilutions: Western Blot: 1:500-1:2500

Type: Primary
Antigen: HEC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty