Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-H2AFX Rabbit Polyclonal Antibody

Supplier: BioVision

3761-100
10004-720EA 252.35 USD InStock
10004-720
Anti-H2AFX Rabbit Polyclonal Antibody
Antibodies
H2AX is a member of the histone H2A family. The four core histones involved in the formation of the nucleosome structure of compacted chromatin are H2A, H2B, H3, and H4. H2AX may function to facilitate DNA repair, and recent studies have shown that H2AX is required for the maintenance of genomic stability.

Recommended Dilutions: Western Blot: 1:500-1:2500; Immunofluorescence: 1:50-1:100

Type: Primary
Antigen: Histone H2AX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty