Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MCAK Rabbit Polyclonal Antibody

Supplier: BioVision

3760-100
10004-718EA 252.35 USD InStock
10004-718
Anti-MCAK Rabbit Polyclonal Antibody
Antibodies
Kinesin activity has been linked to various cellular functions such as vesicle transport, mitotic spindle formation, chromosome segregation, chromosome congression, and cytokinesis. Structurally, all kinesins contain a motor domain with microtubule and nucleotide binding sites that utilize ATP to target cargo along microtubule filaments. MCAK (Mitotic Centromere-Associated Kinesin) is the founding member of kinesin-13 proteins. There are 3 independent genes coding for kinesin-13 proteins kif2a, kif2b, and kif2c (MCAK). Instead of functioning as a translocator of microtubules MCAK is a depolymerizer that is primarily responsible for releasing improper microtubule-kinetochore attachments during cell division.

Recommended Dilutions: Western Blot: 1:500-1:2000

Type: Primary
Antigen: MCAK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty