Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MCAF Rabbit Polyclonal Antibody

Supplier: BioVision

3759-100
10004-716EA 252.35 USD InStock
10004-716
Anti-MCAF Rabbit Polyclonal Antibody
Antibodies
MBD1-Containing Chromatin-Associated Factor (MCAF) associates with methyl CpG-binding protein (MBD1) that functions to recruit methyltransferases such as SETDB1 to chromatin. MCAF is required for the transcriptional repression and heterochromatin formation mediated by MBD1 and SETDB1.

Recommended Dilutions: Western Blot: 1:500-1:2000

Type: Primary
Antigen: MCAF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty