Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-XLF Rabbit Polyclonal Antibody

Supplier: BioVision

3757-100
10004-712EA 252.35 USD InStock
10004-712
Anti-XLF Rabbit Polyclonal Antibody
Antibodies
XLF (also called Cernunnos) interacts with the XRCC4-DNA ligase IV complex to promote DNA non-homologous end-joining. It directly interacts with the XRCC4-Ligase IV complex and siRNA-mediated downregulation of XLF in human cell lines leads to radio-sensitivity and impaired DNA non-homologous end-joining. XLF is homologous to the yeast non-homologous end-joining factor Nej1.

Recommended Dilutions: Western Blot: 1:500-1:2500; Immunoprecipitation

Type: Primary
Antigen: XLF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty