Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-VCP Rabbit Polyclonal Antibody

Supplier: BioVision

3756-100
10004-710EA 272.95 USD InStock
10004-710
Anti-VCP Rabbit Polyclonal Antibody
Antibodies
Valosin containing protein (VCP), also designated TERA (for transitional endoplasmic reticulum ATPase) or p97, is a member of the AAA family of ATPases, which are involved in a variety of cellular activities. VCP is the mammalian homolog of Saccharomyces cerevisiae Cdc48, a protein essential for the completion of mitiosis in yeast. VCP is thought to be involved in a variety of membrane functions and in the regulation of the cell cycle. It associates with ubiquitinated IκB-αas well as with the 26S Proteosome, indicating a potential role for VCP in the proteosome-mediated degradation of IκB-α.

Recommended Dilutions: Western Blot: 1:500-1:2000; immunoprecipitation

Type: Primary
Antigen: VCP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty