Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SMC2 Rabbit Polyclonal Antibody

Supplier: BioVision

3754-100
10004-706EA 252.35 USD InStock
10004-706
Anti-SMC2 Rabbit Polyclonal Antibody
Antibodies
Structural maintenance of chromosomes (SMC) proteins are part of the cohesin and condensin multisubunit complexes that play a critical role in chromosome organization, segregation, gene regulation, and DNA repair. The core condensin complex includes a heterodimer of SMC2 and SMC4 that functions to create positively supercoiled loops for chromatin condensation and metaphase chromatid shaping. A function for condensin in the DNA damage checkpoint response and gene repression has also been suggested.

Recommended Dilutions: Western Blot: 1:500-1:2000

Type: Primary
Antigen: SMC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty