Anti-PI-8 Rabbit Polyclonal Antibody

Supplier: BioVision
3748-100
10004-700EA 286.04 USD
10004-700
Anti-PI-8 Rabbit Polyclonal Antibody
High molecular weight serine protease inhibitors (serpins) are large superfamily of proteins, which bind to and inactivate serine proteinases. In 1995 Sprecher et al. cloned 2 novel serpins, termed PI8 and PI9, from human placenta cDNA library. Sequencing showed that PI8 encodes 376-amino acids. PI8 are localized to the cytoplasm of transfected cells and was able to form an SDS- complex. PI8 is implicated to inhibit furin enzymatic activity and plays a role in platelet-regulated pathophysiological responses.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: PI-8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR