Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-USP7 Rabbit Polyclonal Antibody

Supplier: BioVision

3747-100
10004-698EA 252.35 USD InStock
10004-698
Anti-USP7 Rabbit Polyclonal Antibody
Antibodies
Ubiquitin (Ub) is a highly conserved protein found ubiquitously in eukaryotic organisms. The conjugation of ubiquitin to proteins is an important means to regulate protein activity for many cellular processes by tagging them for degradation. Removal of Ub can rescue proteins from degradation. This is accomplished by the ubiquitin-specific processing protease (UBP) family of enzymes. Ubiquitin specific processing protease 7 (USP7) is a member of this family that has been shown to interact and stabilize p53.

Recommended Dilutions: Western Blot: 1:500-1:2000

Type: Primary
Antigen: USP7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Chicken

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty