Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SLK Rabbit Polyclonal Antibody

Supplier: BioVision

3746-100
10004-696EA 252.35 USD InStock
10004-696
Anti-SLK Rabbit Polyclonal Antibody
Antibodies
SLK (STE20-like kinase), also known as STK2 (serine/threonine protein kinase 2) or se20-9, is a member of the serine/threonine kinase subfamily, Ste20. This subfamily is comprised of several mammalian kinases which exhibit sequence similarity to the Saccharomyces cerevisiae serine/threonine kinase Ste20, a protein involved in relaying signals from G protein-coupled receptors to cytosolic MAP kinase cascades. Members of this subfamily include KHS, GLK, YSK1, HPK1, Krs-1, Krs-2, GC kinase, HGK and SLK. SLK is a ubiquitously expressed protein that localizes to the cytoplasm and contains an N-terminal protein kinase domain, a central coiled-coil domain and a C-terminal ATH domain. SLK is activated through cleavage by caspase-3. SLK indirectly associates with microtubules and plays an important role in cellular stress, cell motility, cell death and cytoskeletal dynamics.

Recommended Dilutions: Western Blot: 1:500-1:2500; immunoprecipitation

Type: Primary
Antigen: SLK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty