Anti-E6AP Rabbit Polyclonal Antibody

Supplier: BioVision
3744-100
10004-692EA 402.79 USD
10004-692
Anti-E6AP Rabbit Polyclonal Antibody
E6AP is an E3 ubiquitin ligase. Inheritance of a mutant maternal copy of this gene results in Angelman syndrome (AS) which is characterized by severe mental retardation, seizures, ataxia, and EEG abnormalities. E6AP interacts with the type 16 and 18 human papillomavirus E6 oncoprotein which results in the targeting of p53 for degradation via the ubiquitin-proteasome system.

Recommended Dilutions: Western Blot: 1:500-1:2500; immunoprecipitation

Type: Primary
Antigen: E6AP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR