The initiation of gene transcription involves the ordered assembly of a multiprotein complex on proximal promoter elements such as the TATA box. In addition to RNA polymerase II, the transcription factors class II (TFII) family of proteins are required for initiation of transcription, as the first step in the formation of this initiation complex is the stable binding of TFIID to the TATA box. An additional TFII related protein, BTF3, does not directly associate with the proximal promoter, but rather forms a stable complex with RNA pol II and facilitates RNA pol II assembling into the complex. The BTF3 gene is ubiquitously expressed and encodes for two protein isoforms, BTF3a and BTF3b, which are produced from alternative splicing. The BTF3 proteins are identical except that BTF3b lacks the first 44 amino acids at the N-terminal of BTF3a. As a consequence of this deletion, BTF3b is unable to induce transcription, despite being able to bind RNA pol II. Additionally, BTF3a and BTF3b associate with the widely expressed protein kinase CK2. CK2 phosphorylates BTF3a, as well as TFIIB, and is required for the efficient transcription of the tRNA and 55 rRNA genes by RNA pol III. BTF3 belongs to the NAC-β family, which includes several related proteins, such as BTF3L1, BTF3L2, BTF3L3 and BTF3L4.
Recommended Dilutions: Western Blot: 1:500-1:2000; immunoprecipitation
Type: Primary
Antigen: BTF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse