Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FOXL2 Rabbit Polyclonal Antibody

Supplier: BioVision

3742-100
10005-912EA 252.35 USD InStock
10005-912
Anti-FOXL2 Rabbit Polyclonal Antibody
Antibodies
FOXL2 is a nuclear protein specially expressed in eyelids and in fetal and adult ovarian follicular cells. Defects in FOXL2 are cause of blepharophimosis, ptosis and epicanthus inversus syndrome (BPES). It is an autosomal dominant disorder characterized by eyelid dysplasia, small palpebral fissures, drooping eyelids and a skin fold. It is also associated with female infertility.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: FoxL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty