Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CTSL Rabbit Polyclonal Antibody

Supplier: BioVision

3741-100
10005-908EA 252.35 USD InStock
10005-908
Anti-CTSL Rabbit Polyclonal Antibody
Antibodies
The cysteine proteases termed lysosomal cathepsins have long been thoμght to be primarily involved in end-stage protein breakdown within lysosomal compartments. Cathepsin L functions to regulate cell cycle progression throμgh proteolytic processing of the CDP/Cux transcription factor.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: CTSL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty