Anti-Tom 40 Rabbit Polyclonal Antibody

Supplier: BioVision
AB273010-100UG
10005-904EA 789.08 USD
10005-904
Anti-Tom 40 Rabbit Polyclonal Antibody
Antibodies
Translocase of the mitochondria Outer Membrane (TOM) is essential for protein import into mitochondria. TOM40 is synthesized in the cytosol and contains information for its mitochondrial targeting and assembly. It is delivered to the mitochondria in an ATP-dependent manner.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: Tom 40
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR