Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BRG1 Rabbit Polyclonal Antibody

Supplier: BioVision

3739-100
10005-902EA 272.95 USD InStock
10005-902
Anti-BRG1 Rabbit Polyclonal Antibody
Antibodies
The acetylation of histone lysine residues plays a crucial role in the epigenetic regulation of gene transcription. A bromodomain is a protein domain that recognizes acetylated lysine residues such as those on the N-terminal tails of histones. This recognition is often a prerequisite for protein-histone association and chromatin remodeling. These domains function in the linking of protein complexes to acetylated nucleosomes, thereby controlling chromatin structure and gene expression. Thus, bromodomains serve as “readers” of histone acetylation marks regulating the transcription of target promoters. BRG1 is a member of the SWI/SNF protein family, which forms part of a large ATP-dependent chromatin remodeling complex. This complex is required for transcriptional activation of genes normally repressed by chromatin. BRG1 is mutated in many cancer cell lines, such as breast, prostate, lung, pancreas and colon. Further, BRG1 has an important role as a tumor suppressor. This protein can be used for the study of bromodomain binding assays, screening inhibitors, and selectivity profiling.

Recommended Dilutions: Western Blot: 1:500-1:2500 µg/ml

Type: Primary
Antigen: BRG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty