Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BRD8 Rabbit Polyclonal Antibody

Supplier: BioVision

3738-100
10005-900EA 272.95 USD InStock
10005-900
Anti-BRD8 Rabbit Polyclonal Antibody
Antibodies
BRD8 interacts with thyroid hormone receptor in a ligand-dependent manner and enhances thyroid hormone-dependent activation from thyroid response elements. BRD8 contains a bromodomain and is thought to be a nuclear receptor coactivator. Three alternatively spliced BRD8 transcript variants that encode distinct BRD8 isoforms have been identified.

Recommended Dilutions: Western Blot: 1:500-1:2000

Type: Primary
Antigen: BRD8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty