Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NIF1 Rabbit Polyclonal Antibody

Supplier: BioVision

3737-100
10005-894EA 272.95 USD InStock
10005-894
Anti-NIF1 Rabbit Polyclonal Antibody
Antibodies
NRC-Interacting Factor 1 (NIF1) interacts with the nuclear receptor transcriptional coactivator NRC and enhances ligand-dependent transcriptional activation by nuclear hormone receptors as well as c-Fos and c-Jun. NIF1 may function to influence chromatin structure.

Recommended Dilutions: Western Blot: 1:500-1:2500;

Type: Primary
Antigen: NIF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty