Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TIP60 Rabbit Polyclonal Antibody

Supplier: BioVision

6126-50
10008-188EA 231.75 USD InStock
10008-188
Anti-TIP60 Rabbit Polyclonal Antibody
Antibodies
TIP60, originally isolated as an HIV-1 TAT-interactive protein, is known to interact with the viral TAT protein that leads to HTATIP polyubiquitination and targets it for degradation. It belongs to the MYST family of histone acetyltransferases (HATs). TIP60 is a catalytic subunit of the NuA4 HAT complex which is involved in transcriptional activation of select genes principally by acetylation of nucleosomal histones H4 and H2A. A co-regulator of transcription factors, TIP60 either promotes or suppresses tumorigenesis. TIP60 is an enzyme that has a role in DNA repair and apoptosis and is thoμght to play an important role in signal transduction.

Recommended Dilutions: Western Blot: 0.25-1.0 µg/ml, IHC (paraffin embedded sections): 5 µg/ml

Type: Primary
Antigen: TIP60
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Chimpanzee, Orangutan, Equine, Canine, Mouse, Bovine, Rat, Opossum, Zebrafish, Xenopus

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty