Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Methyl Lysine Rabbit Polyclonal Antibody (Biotin)

Supplier: BioVision

6124-50
10008-184EA 272.95 USD InStock
10008-184
Anti-Methyl Lysine Rabbit Polyclonal Antibody (Biotin)
Antibodies
Post-translational modifications of proteins play critical roles in the regulation and function of many known biological processes. Proteins can be post-translationally modified in many different ways, and a common post-transcriptional modification of lysine involves methylation. Lysine can be methylated once, twice, or three times by lysine methyltransferases. The transfer of methyl groups from S-adenosyl methionine to histones is catalyzed by enzymes known as histone methyltransferases. Histones which are methylated on certain residues can act epigenetically to repress or activate gene expression.

Recommended Dilutions: Western Blot: 1:2000-1:5000, IP/ELISA: 1:1000

Type: Primary
Antigen: Methyl Lysine
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Multiple Species

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty