Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-EZH1 Rabbit Polyclonal Antibody

Supplier: BioVision

6123-50
10008-182EA 211.15 USD InStock
10008-182
Anti-EZH1 Rabbit Polyclonal Antibody
Antibodies
EZH1 is a human homolog of the Drosophila gene Enhancer of zeste E(z), a member of the polycomb group of transcriptional repressors. In mice, EZH1 is a regulator of homeotic gene expression implicated in the assembly of repressive protein complexes in chromatin. The strong sequence conservation between species sμggests potential role for EZH1 in human development as a transcriptional regulator and as a component of protein complexes that stably maintain heterochromatin.

Recommended Dilutions: Western Blot: 1-2 µg/ml

Type: Primary
Antigen: EZH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty