Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HIST1H3A Mouse Monoclonal Antibody [clone: 117C826]

Supplier: BioVision

6119-50
10008-180EA 252.35 USD InStock
10008-180
Anti-HIST1H3A Mouse Monoclonal Antibody [clone: 117C826]
Antibodies
The phosphorylation of histone H3 plays an important role in gene expression, chromatin remodeling, chromosome condensation, and cell division. Histone H3 phosphorylation may initiate at different phases of the cell division in different organisms, but metaphase chromosomes are always found to be heavily phosphorylated. H3 phosphorylation at serine 28 is coupled with mitotic chromosome condensation in diverse mammalian cell lines.

Recommended Dilutions: Western Blot: 2 µg/ml

Type: Primary
Antigen: HIST1H3A
Clonality: Monoclonal
Clone: 117C826
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty