Anti-DNMT1 Mouse Monoclonal Antibody [clone: 60B1220.1]

Supplier: BioVision
6110-50
10007-994EA 262.69 USD
10007-994
Anti-DNMT1 Mouse Monoclonal Antibody [clone: 60B1220.1]
Methylation of DNA at cytosine residues plays an important role in regulation of gene expression, genomic imprinting, and is essential for mammalian development. Hypermethylation of CpG islands in tumor suppressor genes or hypomethylation of bulk genomic DNA may be linked with development of cancer. To date, three families of mammalian DNA methyltransferase genes have been identified which include DNMT1, DNMT2, and DNMT3. DNMT1 is constitutively expressed in proliferating cells and inactivation of this gene causes global demethylation of genomic DNA and embryonic lethality. DNMT1 co-purifies with the retinoblastoma (Rb) tumour suppressor gene product, E2F1, and HDAC1. DNMT1 also cooperates with Rb to repress transcription from promoters containing E2F-binding sites sμggesting a link between DNA methylation, histone deacetylase, and sequence-specific DNA binding activity, as well as a growth-regulatory pathway that is disrupted in nearly all cancer cells.

Recommended Dilutions: Western Blot: 2-4 µg/ml; ChIP and IP: 1-2 µg/ml

Type: Primary
Antigen: DNMT1
Clonality: Monoclonal
Clone: 60B1220.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR