Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-JMJD6 Rabbit Polyclonal Antibody

Supplier: BioVision

6109-50
10007-992EA 231.75 USD InStock
10007-992
Anti-JMJD6 Rabbit Polyclonal Antibody
Antibodies
Tudor domains are small protein structural motifs of about ~50 amino acids related to the “royal family” of methyl readers, which also includes chromo, MBT, PWWP, and Agenet-like domains. Tudor domains occur either alone, in tandem, or with other domains and are found in many proteins that are involved in RNA metabolism, germ cell development, transposon silencing, DNA damage response, histone modification, and chromatin remodeling. The tudor domains recognize symmetric methylated arginine or methylated lysine residues. JMJD6 is a 403 amino acid nuclear protein lysyl-hydroxylase that has been reported to have arginine demethylase activity for histone H3 at ‘Arg-2’ and histone H4 at ‘Arg-3’. JMJD6 has been sμggested to function in the differentiation of multiple organs during embryogenesis and regulate hematopoietic differentiation and macrophage cytokine responses. First identified as a putative phosphatidylserine receptor involved in phagocytosis of apoptotic cells, JMJD6 was later indicated not to directly function in the clearance of apoptotic cells. It is ubiquitously expressed in most tissues and is upregulated in patients with chronic pancreatitis and upon cytokine treatment, but not upon TNF-α treatment.

Recommended Dilutions: Western Blot: 0.25-1.0 µg/ml

Type: Primary
Antigen: JMJD6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Chimpanzee, Bovine, Canine, Equine, Mouse, Opossum

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty