Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-JMJD2A Rabbit Polyclonal Antibody

Supplier: BioVision

6108-50
10007-990EA 211.15 USD InStock
10007-990
Anti-JMJD2A Rabbit Polyclonal Antibody
Antibodies
Jumonji Domain containing 2A (JMJD2A) is a lysine specific demethylase with emerging roles in histone modification or epigenetic remodeling. This JMJD2A polyclonal antibody was raised against an N-terminal recombinant fragment of JMJD2A. This fragment includes amino acids 1-350 including the JMJN and JMJC domains but not the two LAP/PHD zinc finger or Tudor domains of the 1,064 amino acid protein. JMJD2A is detected at 135 kDa by western blotting using a DLD1 cell lysate as a positive control. Also, the expression of JMJD2A varies by tissue, and northern blotting sμggests skeletal muscle as a negative control and lung as a positive control tissue.

Recommended Dilutions: Western Blot: 1:200 dilution

Type: Primary
Antigen: JMJD2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human Jmjd2A. Expected To Cross React With Jmjd2C (>90% Similarity), Jmjd2B (>85% Similarity) Due To High Homology To These Jmjd2 Paralogs Considering The Antigen Used. Observed Detection Of Purified Recombinant Human Jmjd2D At 50 Ng S?Ggesting Only ~10%

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty