Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SIRT7 Rabbit Polyclonal Antibody

Supplier: BioVision

6107-50
10007-988EA 211.15 USD InStock
10007-988
Anti-SIRT7 Rabbit Polyclonal Antibody
Antibodies
The sirtuins represent a distinct class of trichostatin A-insensitive lysyl-deacetylases (class III HDACs) and have been shown to catalyze a reaction that couples lysine deacetylation to the formation of nicotinamide and O-acetyl-ADP-ribose from NAD+ and the abstracted acetyl group. SIRT7 is a member of this family of proteins and is present in prokaryotes and eukaryotes. All SIR2-like proteins have a sirtuin core domain, which contains a series of sequence motifs conserved in organisms ranging from bacteria to humans. Bacterial, yeast, and mammalian sirtuins are able to metabolize NAD+ and several act as mono-ADP-ribosyltransferases. The enzymatic function of sirtuins is not yet completely understood but as mentioned above, recent reports of histone-activated SIR2-mediated NAD+ metabolism and NAD+-activated SIR2-mediated histone deacetylation sμggest a possible coupled reciprocal activation mechanism involving interactions of SIR2 with NAD+ and the N-ε-acetyl-lysine groups of acetylated histone.

  • Recommended Dilutions: Western Blot: 1-3 µg/ml

Type: Primary
Antigen: SIRT7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty