Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-APP Rabbit Polyclonal Antibody

Supplier: BioVision

6001-100
10007-976EA 272.95 USD InStock
10007-976
Anti-APP Rabbit Polyclonal Antibody
Antibodies
Accumulation of the amyloid-β peptide (Aβ) in the cerebral cortex is a critical event in the pathogenesis of Alzheimer’s disease. The β-amyloid protein precursor (APP) is cleaved by β-secretase, producing a soluble derivative of the protein and a membrane anchored 99-amino acid carboxy-terminal fragment (C99). The C99 fragment serves as substrate for g-secretase to generate the 4 kDa amyloid-β peptide, which is deposited in the brains of all suffers of Alzheimer’s disease.

  • Recommended Dilutions: Western Blot: 1-4 µg/ml; Immunohistochemistry: 2-10 ug/ml

Type: Primary
Antigen: APP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty