Anti-RETN Rabbit Polyclonal Antibody

Supplier: BioVision
5999-100
10007-974EA 286.04 USD
10007-974
Anti-RETN Rabbit Polyclonal Antibody
Resistin (also called adipose tissue-specific secretory factor, ADSF) is a member of the family of adipocyte secreted proteins. Resistin appears to act on skeletal muscle myocytes, hepatocytes, and adipocytes themselves, where it is sμggested to reduce insulin sensitivity leading to type 2 diabetes.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: RETN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR