Anti-RETN Rabbit Polyclonal Antibody

Supplier: BioVision
5997-100
10008-146EA 286.04 USD
10008-146
Anti-RETN Rabbit Polyclonal Antibody
Resistin (also called adipose tissue-specific secretory factor, ADSF) is a member of the family of adipocyte-secreted proteins. Resistin appears to act on skeletal muscle myocytes, hepatocytes, and adipocytes themselves, where it is sμggested to reduce insulin sensitivity leading to type-2 diabetes.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: RETN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR