Anti-C5a Rabbit Polyclonal Antibody

Supplier: BioVision
5995-100
10008-144EA 286.04 USD
10008-144
Anti-C5a Rabbit Polyclonal Antibody
Human complement C5a is a 74-amino acid glycopeptide generated during complement activation from the α-chain of complement C5. C5a is a potent chemotactic factor for human peripheral blood neutrophils and monocytes, and is believed to play an important role in a number of inflammatory conditions.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: C5a
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR